Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serine, USP, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
56-45-1
C3H7NO3
105.09
MTCFGRXMJLQNBG-UHFFFAOYNA-N
2-amino-3-hydroxypropanoic acid
NC(CO)C(O)=O
56-45-1
C3H7NO3
105.09
MTCFGRXMJLQNBG-UHFFFAOYNA-N
2-amino-3-hydroxypropanoic acid
NC(CO)C(O)=O

S1023, 56-45-1, C3H7NO3

Spectrum™ Serine, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Ignition Residue 0.001
Quantity 1 kg
Loss on Drying 0.002
Specific Rotation +14.0° to +15.6°
CAS Max % 1
Percent Purity 98.5 to 101.5%
Grade USP