Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Serotonin Ab-1, Mouse Monoclonal Antibody

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Thermo Scientific™ Serotonin Ab-1, Mouse Monoclonal Antibody.

$357.00 - $507.50

Specifications

Antigen Serotonin Ab-1
Clone 5HT-H209
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MS1431R7
View Documents
Thermo Scientific™
MS1431R7
7mL Each for $357.00
Add to cart
 
MS1431S
View Documents
Thermo Scientific™
MS1431S
1mL Each for $507.50
Add to cart
 
MS1431S1
View Documents
Thermo Scientific™
MS1431S1
500μL Each for $395.50
Add to cart
 
MS1431S0
View Documents
Thermo Scientific™
MS1431S0
100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS1431R1
View Documents
Thermo Scientific™
MS1431R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Thermo Scientific™ Serotonin Ab-1, Mouse Monoclonal Antibody.

Serotonin-positive cells are present in gastric, duodenal, jejunal, ileal, colonic, and appendiceal mucosae. Positive cells are sparse and usually occur in the lower half of the gastric and intestinal mucosae. In the appendix, in addition to the mucosal serotonin-containing cells, there are single or small clusters of serotonin-positive cells in the sub-epithelial region of the lamina propria.

Host Species: Mouse

Clone: 5HT-H209

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: 5-Hydroxytryptamine hydrochloride [3-(2-aminoethyl)-5-hydroxyindole hydrochloride; serotonin hydrochloride]

Positive Control: Appendix

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Serotonin Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG2a kappa
5HT-H209
Monoclonal
Murine
Neuroscience
5-Hydroxytryptamine hydrochloride (3-(2-aminoethyl)-5-hydroxyindole;serotonin)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit