Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serotonin hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number AC215021000
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
153-98-0
C10H12N2O·HCl
212.68
MFCD00012686
MDIGAZPGKJFIAH-UHFFFAOYSA-N
serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride
160436
3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride
C1=CC2=C(C=C1O)C(=CN2)CCN.Cl
153-98-0
C10H12N2O·HCl
212.68
MFCD00012686
MDIGAZPGKJFIAH-UHFFFAOYSA-N
serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride
160436
3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride
C1=CC2=C(C=C1O)C(=CN2)CCN.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 168°C to 172°C
Color Yellow to Beige
Loss on Drying 2.0 % max. (115°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Quantity 100 mg
UN Number 2811
Beilstein 22, III, 5668
Merck Index 14, 8462
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
Formula Weight 212.68
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Serotonin hydrochloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit