Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serotonin hydrochloride, 98%, Acros Organics

Click to view available options
Quantity:
100mg
500mg
Packaging:
Glass bottle
153-98-0
C10H12N2O·HCl
212.68
MFCD00012686
MDIGAZPGKJFIAH-UHFFFAOYSA-N
serotonin hydrochloride,5-hydroxytryptamine hydrochloride,3-2-aminoethyl-1h-indol-5-ol hydrochloride,serotonin hcl,5-hydroxytryptamine hcl,unii-gkn429m9vs,ccris 4420,1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride,gkn429m9vs,3-2-aminoethyl indol-5-ol, chloride
160436
3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride
C1=CC2=C(C=C1O)C(=CN2)CCN.Cl
153-98-0
C10H12N2O·HCl
212.68
MFCD00012686
MDIGAZPGKJFIAH-UHFFFAOYSA-N
serotonin hydrochloride,5-hydroxytryptamine hydrochloride,3-2-aminoethyl-1h-indol-5-ol hydrochloride,serotonin hcl,5-hydroxytryptamine hcl,unii-gkn429m9vs,ccris 4420,1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride,gkn429m9vs,3-2-aminoethyl indol-5-ol, chloride
160436
3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride
C1=CC2=C(C=C1O)C(=CN2)CCN.Cl
Packaging Glass bottle
Melting Point 168°C to 172°C
Color Yellow to Beige
Loss on Drying 2.0 % max. (115°C, 3 hrs) (vacuum)
Sulfated Ash 0.1% max.
Appearance (1% in methanol) clear to slightly hazy, colorless to light yellow
Quantity 500mg
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
UN Number 2811
Beilstein 22, III, 5668
Merck Index 14, 8462
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
Formula Weight 212.68
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Serotonin hydrochloride, 99%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Warning