Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521385
Description
Recommended positive controls: A431. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Pig (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene interacts with target proteins during their translocation into the lumen of the endoplasmic reticulum. Protects unfolded target proteins against degradation during ER stress. May facilitate glycosylation of target proteins after termination of ER stress. May modulate the use of N-glycosylation sites on target proteins.
Specifications
SERP1 | |
Polyclonal | |
Unconjugated | |
SERP1 | |
D3Ucla1; RAMP4; ribosome associated membrane protein 4; ribosome-attached membrane protein 4; Serp1; Stress-associated endoplasmic reticulum protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27230, 28146 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y6X1, Q9Z1W5 | |
SERP1 | |
Synthetic peptide corresponding to a region within amino acids 1 and 51 of SERP1 (Uniprot ID#Q9Y6X1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction