Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin A1/alpha 1-Antitrypsin Rat anti-Mouse, DyLight 550, Clone: RM0527-3E39, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP241750R

Catalog No. NBP241750R


Add to cart

Description

Description

Serpin A1/alpha 1-Antitrypsin Monoclonal antibody specifically detects Serpin A1/alpha 1-Antitrypsin in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Serpin A1/alpha 1-Antitrypsin
RM0527-3E39
Western Blot
Purified
SERPINA1
Recombinant Mouse Serpin A1; aa1-aa413.
Protein G purified
Stem Cell Markers
Primary
5265
Mouse
Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
A1A, AATMGC23330, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Rat
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody recognizes mouse Serpin A1 by direct ELISAs and WBs.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit