Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin A12 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | Serpin A12 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB125167
|
Novus Biologicals
NBP310133100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
Serpin A12 Polyclonal specifically detects Serpin A12 in Mouse samples. It is validated for Western Blot.Specifications
Serpin A12 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
antitrypsin), member 12, member 12, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor | |
The immunogen is a synthetic peptide directed towards the middle region of mouse Serpin A12 (NP_080811.1). Peptide sequence SRLGISKIFEENGDLTRISSHRSLKVGEAVHKAELKMDEKGMEGAAGSGA | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
145264 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title