Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin A3/alpha 1-Antichymotrypsin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$183.00 - $490.00
Specifications
| Antigen | Serpin A3/alpha 1-Antichymotrypsin |
|---|---|
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
Serpin A3/alpha 1-Antichymotrypsin Polyclonal specifically detects Serpin A3/alpha 1-Antichymotrypsin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Serpin A3/alpha 1-Antichymotrypsin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 12 | |
| alpha-1-antichymotrypsin isolated from human serum. | |
| Primary | |
| Store at 4°C. |
| Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer | |
| AACTMGC88254, ACTGIG25, alpha-1-antichymotrypsin, antitrypsin), member 3, Cell growth-inhibiting gene 24/25 protein, growth-inhibiting protein 24, growth-inhibiting protein 25, member 3, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 3, Serpin A3, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) | |
| SERPINA3 | |
| IgG | |
| Affinity purified | |
| The is specific against human alpha-1-antichymotrypsin. This stains monocytes and macrophages in human tissues and does not stain any other cell type. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title