Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin A3/alpha 1-Antichymotrypsin Mouse anti-Human, Alexa Fluor™ 594, Clone: AACT/1451, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254339AF594

Catalog No. NP24339A94


Add to cart

Description

Description

Serpin A3/alpha 1-Antichymotrypsin Monoclonal antibody specifically detects Serpin A3/alpha 1-Antichymotrypsin in Human samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications

Specifications

Serpin A3/alpha 1-Antichymotrypsin
AACT/1451
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
AACTMGC88254, ACTGIG25, alpha-1-antichymotrypsin, antitrypsin), member 3, Cell growth-inhibiting gene 24/25 protein, growth-inhibiting protein 24, growth-inhibiting protein 25, member 3, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 3, Serpin A3, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
SERPINA3
Recombinant human Antichymotrypsin (AACT) protein fragment (aa49-187) (exact sequence is proprietary)
0.1 mL
Primary
12
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit