Learn More
Serpin A4/Kallistatin Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Manufacturer:  R&D Systems AF1669SP
Catalog No. AF1669SP
Add to cart
This item is not returnable. View return policy
Description
Description
Serpin A4/Kallistatin Polyclonal specifically detects Serpin A4/Kallistatin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Neutralization.
Specifications
Specifications
Serpin A4/Kallistatin
LYOPH
Western Blot 0.25 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 10.8 ug/mL
AAA59454
antitrypsin), member 4, KAL, kallistatin, KLST, KSTKallikrein inhibitor, member 4, PI-4, PI4Peptidase inhibitor 4, protease inhibitor 4 (kallistatin), serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A4, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Goat
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Serpin A4/Kallistatin in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, rhSerpin A5, rhSerpin C1, recombinant mouse Serpin D1, and rhSerpin F2 is observed.
Human
Western Blot, Immunohistochemistry, Immunoprecipitation, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SERPINA4
Mouse myeloma cell line NS0-derived recombinant human Serpin A4/Kallistatin Gly20-Pro427 (predicted) Accession # AAA59454
Antigen Affinity-purified
RUO
Primary
5267
Reconstitute at 0.2 mg/mL in sterile PBS.