Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Serpin A8/Angiotensinogen Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

$293.00 - $629.00

Specifications

Target Serpin A8/Angiotensinogen
Conjugate HRP
Target Species Human
Gene ID (Entrez) 183
Gene Symbol AGT
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB009699
View Documents
Novus Biologicals™
NBP27933015PLATES
15 Plates
Each for $629.00
Add to cart
 
NB009700
View Documents
Novus Biologicals™
NBP2793305PLATES
5 Plates
Each for $293.00
Add to cart
 
Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Human Serpin A8/Angiotensinogen.
Specifications

Specifications

Serpin A8/Angiotensinogen
Human
AGT
HRP
183
Rabbit/Mouse
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit