Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin B3/SCCA1 Mouse anti-Human, Clone: CPTC-SERPINB3-2, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | Serpin B3/SCCA1 |
---|---|
Clone | CPTC-SERPINB3-2 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Protein Array |
Applications | Western Blot, Peptide Array |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB123044
|
Novus Biologicals
NBP30778520UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
Serpin B3/SCCA1 Monoclonal specifically detects Serpin B3/SCCA1 in Human samples. It is validated for Western Blot, Protein Array.Specifications
Serpin B3/SCCA1 | |
0.2 mg/ml | |
Western Blot, Peptide Array | |
Unconjugated | |
Mouse | |
Cancer | |
10 mM PBS with 0.05% BSA | |
6317 | |
IgG1 κ | |
Protein A or G purified |
CPTC-SERPINB3-2 | |
Western Blot 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
HsT1196, Protein T4-A, SCC, SCCA, SCCA1SCCA-1, SCCA-PD, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3, serpin B3, serpin peptidase inhibitor, clade B (ovalbumin), member 3, Squamous cell carcinoma antigen 1, T4-A | |
Recombinant human full-length Serpin B3/SCCA1 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title