Learn More
Serpin E1/PAI-1 Mouse anti-Human, Alexa Fluor™ 532, Clone: I-1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223509AF532
Catalog No. NP22309A32
Add to cart
Description
Description
Serpin E1/PAI-1 Monoclonal antibody specifically detects Serpin E1/PAI-1 in Human samples. It is validated for Western Blot,ELISA,Immunohistochemistry.
Specifications
Specifications
Serpin E1/PAI-1
I-1
Western Blot, ELISA, Immunohistochemistry
Endothelial plasminogen activator inhibitor, PAI1PAI-1, PAISerpin E1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Clone I-1 is specific for human PAI-1. No cross-reactivity is observed with PAI-2 as tested by ELISA and immunoblotting. No reactivity is observed with any other human plasma protein when tested by immunoblotting.
ELISA, Immunohistochemistry, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
SERPINE1
Partiallty purified human PAI-1 (6)
0.1 mL
Primary
5054
Human