Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin E2/PN1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220314
Description
Serpin E2/PN1 Polyclonal antibody specifically detects Serpin E2/PN1 in Human, Mouse samples. It is validated for Western Blot.Specifications
Serpin E2/PN1 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2 | |
Rabbit | |
44 kDa | |
0.1 mL | |
Neuroscience | |
5270 | |
Human, Mouse | |
IgG |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
P07093 | |
SERPINE2 | |
Recombinant protein encompassing a sequence within the center region of human SERPINE2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Mouse. Species reactivity based on sequence identity: Human (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction