Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin F2/alpha 2-Antiplasmin Antibody (RM0324-2H63), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211890AF488
Description
Serpin F2/alpha 2-Antiplasmin Monoclonal specifically detects Serpin F2/alpha 2-Antiplasmin in Mouse samples. It is validated for Western Blot.Specifications
Serpin F2/alpha 2-Antiplasmin | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SERPINF2 | |
Mouse recombinant Serpin F2/alpha 2-Antiplasmin | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot | |
RM0324-2H63 | |
Western Blot | |
AAPclade F (alpha-2 antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2 | |
Rat | |
Protein A or G purified | |
Cardiovascular Biology | |
5345 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction