Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin F2/alpha 2-Antiplasmin Mouse anti-Human, Clone: 236122, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
$136.67 - $586.67
Specifications
Antigen | Serpin F2/alpha 2-Antiplasmin |
---|---|
Clone | 236122 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Serpin F2/alpha 2-Antiplasmin Monoclonal specifically detects Serpin F2/alpha 2-Antiplasmin in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Serpin F2/alpha 2-Antiplasmin | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
P08697 | |
5345 | |
Mouse myeloma cell line NS0-derived recombinant human Serpin F2/alpha 2 -Antiplasmin Met28-Lys491 Accession # P08697 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Serpin F2/alpha 2 -Antiplasmin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Serpin A1, A3, A4, A5, recombinant mouse Serpin C1, D1, F2, or recombinant mouse PEDF is observed. |
236122 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AAPclade F (alpha-2 antiplasmin, alpha 2-Antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2 | |
SERPINF2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title