Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Serpin G1/C1 Inhibitor Antibody Pair [HRP]

Monoclonal Antibody Pair

$891.50

Specifications

Target Serpin G1/C1 Inhibitor
Conjugate HRP
Target Species Human
Gene ID (Entrez) 710
Gene Symbol SERPING1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB009701
View Documents
Novus Biologicals™
NBP27933115PLATES
15 Plates
Each for $891.50
Add to cart
 
NB009702
View Documents
Novus Biologicals™
NBP2793315PLATES
5 Plates This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Serpin G1/C1 Inhibitor.
Specifications

Specifications

Serpin G1/C1 Inhibitor
Human
SERPING1
HRP
710
Rabbit/Mouse
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit