Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin G1/C1 Inhibitor Mouse anti-Human, Clone: 8 F3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105156
Description
Serpin G1/C1 Inhibitor Monoclonal specifically detects Serpin G1/C1 Inhibitor in Human samples. It is validated for ELISA.Specifications
Serpin G1/C1 Inhibitor | |
Monoclonal | |
1.0 mg/mL | |
ELISA 1:16000 | |
C1IN, C1INH, C1NH, HAE1, HAE2, SERPING1 serpin peptidase inhibitor, clade G (C1 inhibitor), member 1 | |
Mouse | |
Protein A or G purified | |
RUO | |
710 | |
Human | |
Purified |
ELISA | |
8 F3 | |
Unconjugated | |
P05155 | |
SERPING1 | |
C1-inhibitor (C1INH) isolated from human plasma | |
100 μg | |
Primary | |
C1 inhibitor | |
Store at 4C. Do not freeze. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only