Learn More
Serpin G1/C1 Inhibitor Rabbit anti-Human, Alexa Fluor 350, Clone: 18, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP289738AF350
Catalog No. NB109023
Add to cart
Description
Description
Serpin G1/C1 Inhibitor Monoclonal specifically detects Serpin G1/C1 Inhibitor in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA Detection.
Specifications
Specifications
Serpin G1/C1 Inhibitor
18
50 mM Sodium Borate
C1IN, C1INH, C1NH, HAE1, HAE2, SERPING1 serpin peptidase inhibitor, clade G (C1 inhibitor), member 1
This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin G1/C1 Inhibitor (NP_000053.2; Met 1-Ala 500).
Protein A purified
RUO
Primary
710
Western Blot, ELISA
Alexa Fluor 350
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human