Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINA1 Goat anti-Human, Polyclonal , Abnova™

Goat polyclonal antibody raised against native human SERPINA1.

Manufacturer:  Abnova Corporation PAB29770

Catalog No. 89-934-918


Add to cart

Description

Description

The protein encoded by this gene is secreted and is a serine protease inhibitor whose targets include elastase, plasmin, thrombin, trypsin, chymotrypsin, and plasminogen activator. Defects in this gene can cause emphysema or liver disease. Several transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Specifications

Specifications

SERPINA1
Unconjugated
ELISA (2 ug/mL) Immunoprecipitation (2 ug/mg) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user.
P01009
A1A/A1AT/AAT/MGC23330/MGC9222/PI/PI1/PRO2275
Goat
IgG
1 mg
Store at 4°C.
Polyclonal
Human
ELISA, Immunoprecipitation, Western Blot
Goat polyclonal antibody raised against native human SERPINA1.
In PBS, pH 7.2 (0.09% sodium azide)
Liquid
SERPINA1
Native purified human SERPINA1.
Antigen Affinity Purified
RUO
Primary
5265
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit