Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINA3 Mouse anti-Human, Clone: AACT/1452, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14479

Catalog No. 89-034-562


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human SERPINA3.

  • High specificity
  • High reactivity
Specifications

Specifications

SERPINA3
AACT/1452
Mouse monoclonal antibody raised against partial recombinant human SERPINA3.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
SERPINA3
Recombinant protein corresponding to amino acids 49-187 of human SERPINA3.
Protein A/G purification
RUO
Primary
12
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined
P01011
AACT|ACT|GIG24|GIG25|MGC88254
Mouse
IgG1
100 ug
Store at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)