Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$150.20 - $425.50
Specifications
Antigen | SERPINA3 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5122247
![]() |
Thermo Scientific
PA5122247 |
100 μL |
Each for $425.50
|
|
|||||
PIPA5122248
![]() |
Thermo Scientific
PA5122248 |
20 μL |
Each for $150.20
|
|
|||||
Description
SERPINA3 Polyclonal antibody specifically detects SERPINA3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
SERPINA3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P01011 | |
12 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300 | |
AACT; ACT; alpha-1-antichymotrypsin; Alpha-1-antichymotrypsin His-Pro-less; Cell growth-inhibiting gene 24/25 protein; GIG24; GIG25; Growth inhibiting protein 24; Growth inhibiting protein 25; growth-inhibiting protein 24; growth-inhibiting protein 25; MGC88254; Serine (or cysteine) proteinase inhibitor clade A member 3; serine (or cysteine) proteinase inhibitor, clade A, member 3; serpin A3; serpin family A member 3; Serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3; SERPINA3 | |
Lys235-Arg394 (Accession P01011), with N-terminal His and GST Tag | |
Primary | |
-20°C | |
SERPINA3 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title