Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINA5 Mouse anti-Human, Clone: OTI2D10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525897

Catalog No. PIMA525897


Add to cart

Description

Description

This gene encodes a member of the serine proteinase inhibitor (serpin) superfamily. This member is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), and hence is an inhibitor of fibrinolysis. Defects in this gene are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency), and high concentrations of the gene product are associated with thrombophilia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

SERPINA5
Monoclonal
0.75 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P05154
PAI-3, PAI3, PCI, PLANH3, PROCI, acrosomal serine protease inhibitor, plasma serine protease inhibitor, plasminogen activator inhibitor 3, plasminogen activator inhibitor III, plasminogen activator inhibitor-3, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitr
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5104
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2D10
Unconjugated
SERPINA5
Liquid
SERPINA5
Full length human recombit protein of SERPINA5 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit