Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINA9 Mouse anti-Human, Clone: 136CT29.7.4, Abnova™

Mouse monoclonal antibody raised against synthetic peptide of human SERPINA9.

Manufacturer:  Abnova Corporation MAB12289

Catalog No. 89-934-919


Add to cart

Description

Description

Specifications

Specifications

SERPINA9
136CT29.7.4
Mouse monoclonal antibody raised against synthetic peptide of human SERPINA9.
In ascites (0.09% sodium azide)
GCET1/MGC131906/SERPINA11/SERPINA11b
Mouse
IgM, kappa
RUO
Primary
327657
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (1:50-1:100) Western Blot (1:1000-1:3000) The optimal working dilution should be determined by the end user.
Liquid
SERPINA9
A synthetic peptide (conjugated with KLH) corresponding to amino acids 158-186 at N-terminus region of human SERPINA9.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit