Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERPINB11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550876
Description
The antibody detects endogenous levels of total SERPINB11 protein.
SerpinB11 (serpin peptidase inhibitor, clade B (ovalbumin), member 11), also known as EPIPIN or SERPIN11, is a 392 amino acid cytoplasmic protein that belongs to the Ov-serpin subfamily and serpin family. Like other members of the serpin family, SerpinB11 has been identified as a noninhibitory intracellular protein. The gene encoding SerpinB11 maps to human chromosome 18, which houses over 300 protein-coding genes and contains nearly 76 million bases. There are a variety of diseases associated with defects in chromosome 18-localized genes, some of which include Trisomy 18 (also known as Edwards syndrome), Niemann-Pick disease, hereditary hemorrhagic telangiectasia, erythropoietic protoporphyria and follicular lymphomas.
Specifications
SERPINB11 | |
Polyclonal | |
Unconjugated | |
SERPINB11 | |
2310046M08Rik; AU015399; EPIPIN; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 11; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 11; serpin B11; serpin family B member 11 (gene/pseudogene); serpin peptidase inhibitor, clade B (ovalbumin), member 11; SERPIN11; SERPINB11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
66957, 89778 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q96P15, Q9CQV3 | |
SERPINB11 | |
Fusion protein of human serpin peptidase inhibitor, clade B (ovalbumin), member 11 (gene/pseudogene). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction