Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB11 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548992

Catalog No. PA548992

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Has no serine protease inhibitory activity, probably due to mutations in the scaffold impairing conformational change.
Specifications

Specifications

SERPINB11
Polyclonal
PBS with 0.09% sodium azide
Q96P15
Serpin B11, SERPINB11
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
89778
Western Blot
Unconjugated
SERPINB11
Liquid
SERPINB11
KLH conjugated synthetic peptide between 170-196 amino acids from the Central region of human SERPINB11.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit