Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERPINB12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101876
Description
Antibody detects endogenous levels of total SERPINB12.
Members of the human serpin family regulate a diverse array of serine and cysteine proteinases associated with essential biological processes such as fibrinolysis, coagulation, inflammation, cell mobility, cellular differentiation, and apoptosis. Most serpins are secreted and attain physiologic concentrations in the blood and extracellular fluids. However, a subset of the serpin superfamily, the ov-serpins, also resides intracellularly. A novel member of the human ov-serpin gene family, SERPINB12 is expressed in many tissues, including brain, bone marrow, lymph node, heart, lung, liver, pancreas, testis, ovary, and intestines. SERPINB12 is an inhibitor of trypsin-like serine proteinases and a new functional member of the human ov-serpin family.
Specifications
SERPINB12 | |
Polyclonal | |
Unconjugated | |
SERPINB12 | |
MGC119247; MGC119248; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 12; serpin B12; serpin family B member 12; serpin peptidase inhibitor, clade B (ovalbumin), member 12; SERPINB12; YUKOPIN | |
Rabbit | |
Affinity chromatography | |
RUO | |
89777 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96P63 | |
SERPINB12 | |
A synthesized peptide derived from human SERPINB12(Accession Q96P63), corresponding to amino acid residues D55-R105. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction