Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB13 Mouse anti-Human, Mouse, Clone: OTI1D4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525731

Catalog No. PIMA525731


Add to cart

Description

Description

Members of the human serpin family regulate a diverse array of serine and cysteine proteinases associated with essential biological processes such as fibrinolysis, coagulation, inflammation, cell mobility, cellular differentiation, and apoptosis. Most serpins are secreted and attain physiologic concentrations in the blood and extracellular fluids. However, a subset of the serpin superfamily, the ov-serpins, also resides intracellularly. Differential display was used to identify a novel serpin (headpin or SERPINB13) that is underexpressed in squamous cell cancers of the oral cavity. SERPINB13 is expressed in normal oral mucosal tissue, skin, and cultured keratinocytes.
Specifications

Specifications

SERPINB13
Monoclonal
0.74 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CDC0, Q9UIV8
Hurpin, HaCaT UV-repressible serpin, Protease inhibitor 13, Headpin
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
241196, 5275
Flow Cytometry, Western Blot
OTI1D4
Unconjugated
SERPINB13
Liquid
SERPINB13
Full length human recombit protein of SERPINB13 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit