Learn More
SerpinB2 Mouse anti-Human, Alexa Fluor 750, Clone: 930109, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8550S100UG
Catalog No. FAB8550S100
Add to cart
This item is not returnable. View return policy
Description
Description
SerpinB2 Monoclonal antibody specifically detects SerpinB2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
SerpinB2
Monoclonal
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
HsT1201, Monocyte Arg-serpin, PAI, PAI2, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2, Urokinase Inhibitor
E. coli-derived recombinant human Serpin B2 Glu2-Ala65, Ala99-Pro415 Accession # P05120
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human Serpin B2 in direct ELISAs.
Flow Cytometry
930109
Flow Cytometry 0.25-1 ug/10^6 cells
P05120
Mouse
IgG2b
RUO
Primary
5055
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)