Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB2 Mouse anti-Human, Clone: OTI1G3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503926

Catalog No. 50-167-4704


Add to cart

Description

Description

PAI2 inhibits urokinase-type plasminogen activator. The monocyte derived PAI-2 is distinct from the endothelial cell-derived PAI-1.
Specifications

Specifications

SERPINB2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P05120
HsT1201, PAI, PAI-2, PAI2, PLANH2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5055
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI1G3
Unconjugated
SERPINB2
Liquid
SERPINB2
Full length human recombit protein of human SERPINB2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit