Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB3/SERPINB4 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

$150.20 - $425.50

Specifications

Antigen SERPINB3/SERPINB4
Concentration 0.5 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5121616
Encompass
View Documents
Thermo Scientific
PA5121616
100 μL
Each for $425.50
Add to Cart
 
PIPA5121617
Encompass
View Documents
Thermo Scientific
PA5121617
20 μL
Each for $150.20
Add to Cart
 
Description

Description

SERPINB3/SERPINB4 Polyclonal antibody specifically detects SERPINB3/SERPINB4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

SERPINB3/SERPINB4
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human
PBS with 50% glycerol and 0.05% Proclin 300
Serpinb3, SERPINB4
Primary
-20°C
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% Proclin 300
P29508, P48594
IgG
Antigen affinity chromatography
Antibody
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit