Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINB6 Mouse anti-Human, Mouse, Clone: OTI1C5, liquid, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene TA504127
Description
SERPINB6 may be involved in the regulation of serine proteinases present in the brain or extravasated from the blood. Inhibitor of cathepsin G, kallikrein-8 and thrombin. SERPINB6 may play an important role in the inner ear in the protection against leakage of lysosomal content during stress and loss of this protection results in cell death and sensorineural hearing loss.Specifications
SERPINB6 | |
Monoclonal | |
0.71 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P35237 | |
SERPINB6, Serpinb6a | |
Full length human recombit protein of human SERPINB6 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1C5 | |
Unconjugated | |
SERPINB6 | |
CAP, DFNB91, MSTP057, PI-6, PI6, PTI, SPI3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
20719, 5269 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction