Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB6 Mouse anti-Human, Mouse, Clone: OTI3H3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504355

Catalog No. 50-167-5070


Add to cart

Description

Description

SERPINB6 may be involved in the regulation of serine proteinases present in the brain or extravasated from the blood. Inhibitor of cathepsin G, kallikrein-8 and thrombin. SERPINB6 may play an important role in the inner ear in the protection against leakage of lysosomal content during stress and loss of this protection results in cell death and sensorineural hearing loss.
Specifications

Specifications

SERPINB6
Monoclonal
1.08 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P35237
CAP, DFNB91, MSTP057, PI-6, PI6, PTI, SPI3
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
20719, 5269
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI3H3
Unconjugated
SERPINB6
Liquid
SERPINB6, Serpinb6a
Full length human recombit protein of human SERPINB6 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit