Learn More
SERPINB6 Mouse anti-Human, Mouse, Non-human primate, Clone: OTI3H3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525889
Catalog No. PIMA525889
Add to cart
Description
Description
The protein encoded by this gene is a member of the serpin superfamily, and ovalbumin-serpin subfamily. It was originally discovered as a placental thrombin inhibitor. The mouse homolog was found to be expressed in the hair cells of the inner ear. Mutations in this gene are associated with nonsyndromic progressive hearing loss, suggesting that this serpin plays an important role in the inner ear in the protection against leakage of lysosomal content during stress, and that loss of this protection results in cell death and sensorineural hearing loss. Alternatively spliced transcript variants have been found for this gene.
Specifications
Specifications
SERPINB6
Monoclonal
1.08 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q60854, Q4R3G2, P35237
SERPINB6 ; serpin peptidase inhibitor, clade B (ovalbumin), member 6 ; CAP; DKFZp686I04222; MGC111370; MSTP057; PI6; PTI; protease inhibitor 6 (placental thrombin inhibitor); serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 6
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
101866493, 20719, 5269
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI3H3
Unconjugated
SERPINB6
Liquid
SERPINB6, Serpinb6a
Full length human recombit protein of SERPINB6 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Non-human primate