Learn More
SERPINB9 Rabbit anti-Human, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group Inc 149101AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-172-9827
Add to cart
Description
Description
PI9 belongs to the large superfamily of serine proteinase inhibitors (serpins), which bind to and inactivate serine proteinases. These interactions are involved in many cellular processes, including coagulation, fibrinolysis, complement fixation, matrix remodeling, and apoptosis (Sprecher et al., 1995 ).
Specifications
Specifications
SERPINB9
Polyclonal
Unconjugated
SERPINB9
CAP 3, CAP3, Cytoplasmic antiproteinase 3, Peptidase inhibitor 9, PI 9, PI9, Serpin B9, SERPINB9
Rabbit
Antigen Affinity Chromatography
RUO
5272
-20°C
Liquid
Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P50453
SERPINB9
SERPINB9 Fusion Protein Ag6707
150 μL
Primary
Human
Antibody
IgG