Learn More
SERPINB9 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$459.80
Specifications
Antigen | SERPINB9 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9827
![]() ![]() |
Proteintech Group Inc
149101AP150UL |
150 μL |
Each of 1 for $459.80
|
|
|||||
Description
PI9 belongs to the large superfamily of serine proteinase inhibitors (serpins), which bind to and inactivate serine proteinases. These interactions are involved in many cellular processes, including coagulation, fibrinolysis, complement fixation, matrix remodeling, and apoptosis (Sprecher et al., 1995 ).Specifications
SERPINB9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P50453 | |
5272 | |
SERPINB9 Fusion Protein Ag6707 | |
Primary | |
-20°C | |
SERPINB9 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CAP 3, CAP3, Cytoplasmic antiproteinase 3, Peptidase inhibitor 9, PI 9, PI9, Serpin B9, SERPINB9 | |
SERPINB9 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.