Learn More
SerpinE1/PAI-1 Mouse anti-Human, Janelia Fluor 549, Clone: I-2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223499JF549
Catalog No. NBP23499549
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
SerpinE1/PAI-1 Monoclonal antibody specifically detects SerpinE1/PAI-1 in Human samples. It is validated for Immunohistochemistry.
Specifications
Specifications
SerpinE1/PAI-1
I-2
Immunohistochemistry
Purified
SERPINE1
Partiallty purified human PAI-1
Protein A or G purified
Cancer
Primary
5054
Human
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
Endothelial plasminogen activator inhibitor, PAI1PAI-1, PAISerpin E1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
Clone I-2 is specific for human PAI-1. No cross-reactivity is observed with PAI-2 as tested by ELISA, immunoblotting and neutralization of inhibitory activity. No reactivity is observed with any other human plasma protein when tested by immunoblotting.