Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINE2 Mouse anti-Human, Clone: 2C9, Abnova™

Mouse monoclonal antibody raised against full length recombinant human SERPINE2.

Manufacturer:  Abnova Corporation MAB12886

Catalog No. 89-934-922


Add to cart

Description

Description

Specifications

Specifications

SERPINE2
2C9
Mouse monoclonal antibody raised against full length recombinant human SERPINE2.
In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide).
GDN/PI7/PN1/PNI
Mouse
IgG1
50 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Liquid
SERPINE2
Recombinant protein corresponding to full length human SERPINE2.
Protein A/G purification
RUO
Primary
5270
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit