Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINE2 Mouse anti-Human, Clone: OTI3B12, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504602

Catalog No. 50-167-5255


Add to cart

Description

Description

SERPINE2 encodes a member of the serpin family of proteins, a group of proteins that inhibit serine proteases. Thrombin, urokinase, plasmin and trypsin are among the proteases that this family member can inhibit. This gene is a susceptibility gene for chronic obstructive pulmonary disease and for emphysema. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

SERPINE2
Monoclonal
0.66 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P07093
GDN, GDNPF, PI-7, PI7, PN-1, PN1, PNI
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5270
Flow Cytometry, Western Blot
OTI3B12
Unconjugated
SERPINE2
Liquid
SERPINE2
Full length human recombit protein of human SERPINE2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit