Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
SERPINE2 Mouse anti-Human, Non-human primate, Clone: OTI6F3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525961
Catalog No. PIMA525961
Description
Description
This gene encodes a member of the serpin family of proteins, a group of proteins that inhibit serine proteases. Thrombin, urokinase, plasmin and trypsin are among the proteases that this family member can inhibit. This gene is a susceptibility gene for chronic obstructive pulmonary disease and for emphysema. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
SERPINE2
Monoclonal
1.09 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P07093
GDN;GDN_HUMAN;Glia derived nexin;Glia-derived nexin;P17;Peptidase inhibitor 7;Pi-7;Plasminogen activator inhibitor type 1, member 2;PN-1;PN1;PNI;Protease inhibitor 7;Protease nexin 1;Protease nexin I;Serpin E2;Serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2;SERPINE 2;Serpine2;
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5270, 707121
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI6F3
Unconjugated
SERPINE2
Liquid
SERPINE2
Full length human recombit protein of SERPINE2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Non-human primate