Learn More
Aviva Systems Biology Corp SERPINH1 Antibody - C-terminal region (AVARP09002_T100) | 100uL
Supplier:  Aviva Systems Biology Corp AVARP09002_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SERPINH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SERPINH1. Alternate names: CBP1, CBP2, OI10, gp46, AsTP3, HSP47, PIG14, PPROM, RA-A47, SERPINH2ApplicationIP, WB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-62559