Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINH1 Mouse anti-Human, Rat, Clone: 134CT3.1.1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524713

Catalog No. PIMA524713

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

SERPINH1 is a member of the serpin superfamily of serine proteinase inhibitors, and its expression is induced by heat shock. The SERPINH1 protein localizes to the endoplasmic reticulum lumen and binds collagen; thus it is thought to be a molecular chaperone involved in the maturation of collagen molecules. Autoantibodies to SERPINH1 have been found in patients with rheumatoid arthritis.
Specifications

Specifications

SERPINH1
Monoclonal
0.3 mg/mL
PBS with 0.09% sodium azide
P29457, P50454
Serpin H1, 47 kDa heat shock protein, Collagen-binding protein, Colligin, GP46, Serpinh1, Cbp1, Hsp47
Mouse
IgG1
400 μL
-20°C
Primary
29345, 871
Immunohistochemistry (Paraffin), Western Blot
134CT3.1.1
Unconjugated
SERPINH1
Liquid
SERPINH1
KLH conjugated synthetic peptide between 222-251 amino acids from the Central region of rat Serpinh1
Protein G
RUO
Antibody
Monoclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit