Learn More
Aviva Systems Biology Corp SERTAD2 Antibody - N-terminal region (ARP51655_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51655_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SERTAD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SERTAD2. Alternate names: MGC126688, MGC126690, Sei-2, TRIP-Br2ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60578