Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serum Amyloid A1/A2 Mouse anti-Human, Clone: 924912, R&D Systems™

Mouse Monoclonal Antibody

$116.82 - $341.00

Specifications

Antigen Serum Amyloid A1/A2
Clone 924912
Dilution ELISA
Applications ELISA
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB30193100 View Documents R&D Systems
MAB30193100
100 ug Each for $341.00
Add to cart
 
MAB30193SP View Documents R&D Systems
MAB30193SP
25 ug Each for $116.82
Add to cart
 
Description

Description

Serum Amyloid A1/A2 Monoclonal antibody specifically detects Serum Amyloid A1/A2 in Human samples. It is validated for ELISA.
Specifications

Specifications

Serum Amyloid A1/A2
ELISA
Unconjugated
Mouse
Human
P0DJI8
E. coli-derived recombinant human Serum Amyloid A1 Arg19-Tyr122 Accession # P0DJI8
Monoclonal
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
924912
ELISA
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
6288
IgG2b
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Serum Amyloid A1 and Serum Amyloid A2 in direct ELISAs.

RUO

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit