Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serum Amyloid A1/A2 Mouse anti-Human, Clone: SAA/326, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Serum Amyloid A1/A2
Clone SAA/326
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123005
View Documents
Novus Biologicals
NBP30776610UG
100 μg
Each for $352.89
Add to cart
 
NB123006
View Documents
Novus Biologicals
NBP30776620UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Serum Amyloid A1/A2 Monoclonal antibody specifically detects Serum Amyloid A1/A2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Serum Amyloid A1/A2
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
Recombinant full-length human Serum Amyloid A1/A2 (SAA) protein
Monoclonal
Protein A or G purified
SAA/326
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Human
6288
IgG2a Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit