Learn More
Greiner Bio-One Serum No Additive Tubes
With non-ridged pull caps
Supplier: Greiner Bio-One 454240
Description
- Use for discard tubes, immunohematology or viral marker testing
- Clear as glass with equal or better performance
- Molded to be virtually unbreakable, providing a better level of safety than glass
- Tubes have an 18-month shelf life from the date of manufacture
- Nonscrew Safety Pull Caps can be removed with a gentle pull
Specifications
No-additive Tubes | |
Non-ridged Pull Cap | |
13 x 75 mm |
Plastic | |
Blue with Black Ring |