Learn More
Cell Biolabs IncSupplier Diversity Partner SERUM TRIGLYCERIDE KIT COLOR  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA396
The Serum Triglyceride Quantification Kit measures triglyceride concentrations in serum, plasma, or lysates by a coupled enzymatic reaction system. Results read on a standard microplate reader. 100 wells including standards and unknowns.
Catalog No. 50-673-582