Servus™ Economy PVC Knee Boots

Graduated boot heights

Manufacturer: Honeywell Safety Products 18822BLM070

 View more versions of this product

Catalog No. 19-019500

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM F2413-11 M I/75 C/75
    For Use With (Application) Agriculture, construction, food services, municipal services, oil and Gas, pharmaceutical, transportation, utilities
    Gender Male
    Height (English) 16 in.
    Seam Type Seamless
    Shank Material Steel
    Sole Features Unique trac10™ outsole, contour cushion insoles
    Toe Style Plain
    Waterproof Yes
    Color Black
    Material PVC
    Size 7

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit