Learn More
Servus™ Economy PVC Knee Boots
Graduated boot heights
Manufacturer:  Honeywell Safety Products 18822BLM080
 View more versions of this product
Catalog No. 19-019501
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
PVC
ASTM F2413-11 M I/75 C/75
Male
Seamless
Unique trac10™ outsole, contour cushion insoles
Yes
8
Agriculture, construction, food services, municipal services, oil and Gas, pharmaceutical, transportation, utilities
16 in.
Steel
Plain
Black