Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Servus™ Injection-Molded Safety Boots, 15 in.

Reinforced construction provides a durable and long lasting boot without sacrificing comfort

Manufacturer:  Honeywell Safety Products 73505BLM110

 View more versions of this product

Catalog No. 19-019-541


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

PVC
15 in.
Black
11
38cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit